Ethyl (2E)-3-{1-methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrol-2-yl}prop-2-enoate ≥95%

Supplier: Matrix Scientific

070785-500MG 070785-1G
101946-320EA 403.08 USD
101946-320 101946-322
Ethyl (2E)-3-{1-methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrol-2-yl}prop-2-enoate ≥95%
Ethyl (2E)-3-{1-methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrol-2-yl}prop-2-enoate

Hazard: Irritant

Formula: C₁₈H₁₉NO₃
Melting Pt: 94…96 °C
MDL Number: MFCD21333202
CAS Number: 428872-08-6

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR