Ethyl-4-chloro-6-methoxy-5-nitropyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

070784-500MG 070784-1G
101946-316EA 803.08 USD
101946-316 101946-318
Ethyl-4-chloro-6-methoxy-5-nitropyridine-3-carboxylate ≥97%
Ethyl-4-chloro-6-methoxy-5-nitropyridine-3-carboxylate

Hazard: Irritant

Formula: C₉H₉ClN₂O₅
Melting Pt: 85…86 °C
MDL Number: MFCD21333211
CAS Number: 1210835-76-9

Order Now

Specification Test Results

Melting point 85-86°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR