1-(2,6-Dichloropyridin-3-yl)ethanone ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Acetyl-2,6-dichloropyridine

070781-500MG 070781-1G
101946-304EA 600 USD
101946-304 101946-306
1-(2,6-Dichloropyridin-3-yl)ethanone ≥97%
1-(2,6-Dichloropyridin-3-yl)ethanone

Hazard: Irritant

Formula: C₇H₅Cl₂NO
MW: 190.0267 g/mol
Melting Pt: 32…34 °C
MDL Number: MFCD11847148
CAS Number: 412018-50-9

Order Now

Specification Test Results

Melting point 32-34°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR