2,6-Bis(trifluoromethyl)pyridine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Bis(trifluoromethyl)isonicotinic acid

070765-500MG 070765-1G
101946-254EA 1593.85 USD
101946-254 101946-256
2,6-Bis(trifluoromethyl)pyridine-4-carboxylic acid ≥95%
2,6-Bis(trifluoromethyl)pyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₈H₃F₆NO₂
MW: 259.11 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD11501683
CAS Number: 1092343-70-8

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR