Benzyl-4-(cyanomethylidene)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

070764-500MG 070764-1G 070764-5G
101946-248EA 298.46 USD
101946-248 101946-250 101946-252
Benzyl-4-(cyanomethylidene)piperidine-1-carboxylate ≥95%
Benzyl-4-(cyanomethylidene)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MW: 256.31 g/mol
Melting Pt: 72…73 °C
MDL Number: MFCD09953297
CAS Number: 1357147-42-2

Order Now

Specification Test Results

Melting point 72-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR