5-(Trifluoromethyl)-1H-pyrazol-3-amine

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-(trifluoromethyl)-1H-pyrazole

067215-500MG 067215-1G
101933-562EA 193.85 USD
101933-562 101933-564
5-(Trifluoromethyl)-1H-pyrazol-3-amine
5-(Trifluoromethyl)-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₄H₄F₃N₃
MW: 151.09 g/mol
Melting Pt: 96…99 °C
MDL Number: MFCD09834152
CAS Number: 852443-61-9

Order Now

Specification Test Results

Melting point 96-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR