tert-Butyl-4-(3,5-dibromopyridin-2-yl)piperazine-1-carboxylate

Supplier: Matrix Scientific

067214-500MG 067214-1G 067214-5G
101933-556EA 193.85 USD
101933-556 101933-558 101933-560
tert-Butyl-4-(3,5-dibromopyridin-2-yl)piperazine-1-carboxylate
tert-Butyl-4-(3,5-dibromopyridin-2-yl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₉Br₂N₃O₂
MW: 421.14 g/mol
Melting Pt: 68…71 °C
MDL Number: MFCD20921612
CAS Number: 1223432-03-8

Order Now

Specification Test Results

Melting point 68-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR