tert-Butyl-5-oxo-7-phenyl-1,4-diazepane-1-carboxylate

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-oxo-7-phenylhomopiperazine-1-carboxylate

067213-500MG 067213-1G
101933-552EA 1192.31 USD
101933-552 101933-554
tert-Butyl-5-oxo-7-phenyl-1,4-diazepane-1-carboxylate
tert-Butyl-5-oxo-7-phenyl-1,4-diazepane-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₃
MW: 290.37 g/mol
Melting Pt: 41…43 °C
MDL Number: MFCD20921596
CAS Number: 220898-16-8

Order Now

Specification Test Results

Melting point 41-43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR