6-tert-Butyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Supplier: Matrix Scientific

Synonyms: 6-(tert-Butyl)-2-hydroxynicotinonitrile

067212-500MG 067212-1G
101933-548EA 600 USD
101933-548 101933-550
6-tert-Butyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
6-tert-Butyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Melting Pt: 194…196 °C
MDL Number: MFCD10478931
CAS Number: 4138-19-6

Order Now

Specification Test Results

Melting point 194-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR