tert-Butyl-N-(3-iodo-5-nitrophenyl)carbamate

Supplier: Matrix Scientific

067211-500MG 067211-1G
101933-544EA 803.08 USD
101933-544 101933-546
tert-Butyl-N-(3-iodo-5-nitrophenyl)carbamate
tert-Butyl-N-(3-iodo-5-nitrophenyl)carbamate

Hazard: Irritant

Formula: C₁₁H₁₃IN₂O₄
Melting Pt: 113…115 °C
MDL Number: MFCD20921570
CAS Number: 1353878-19-9

Order Now

Specification Test Results

Melting point 113-115°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR