tert-Butyl-4-(6-chloropyridin-2-yl)piperazine-1-carboxylate

Supplier: Matrix Scientific

067207-500MG 067207-1G
101933-528EA 403.08 USD
101933-528 101933-530
tert-Butyl-4-(6-chloropyridin-2-yl)piperazine-1-carboxylate
tert-Butyl-4-(6-chloropyridin-2-yl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₀ClN₃O₂
MW: 297.79 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD18447576
CAS Number: 1239369-20-0

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR