1H-Pyrazolo[3,4-b]pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
7-Azaindazole, Pyrazolo[3,4-b]pyridine
067203-1G
067203-5G
101933-512EA
27.69
USD
101933-512
101933-514
1H-Pyrazolo[3,4-b]pyridine ≥95%
1H-Pyrazolo[3,4-b]pyridine
Hazard: Irritant
Formula:
C₆H₅N₃ MW: 119.12 g/mol Melting Pt: 93…95 °C |
MDL Number:
MFCD05663981 CAS Number: 271-73-8 |
Specification Test Results
Melting point | 93-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...