7-Phenyl-1,4-diazepan-5-one

Supplier: Matrix Scientific

Synonyms: 7-Phenylhomopiperazin-5-one

067201-500MG 067201-1G
101933-502EA 803.08 USD
101933-502 101933-504
7-Phenyl-1,4-diazepan-5-one
7-Phenyl-1,4-diazepan-5-one

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MW: 190.25 g/mol
Melting Pt: 104…106 °C
MDL Number: MFCD12197004
CAS Number: 89044-79-1

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR