Methyl-6-cyano-3-methoxy-2-methylbenzoate

Supplier: Matrix Scientific

067198-500MG 067198-1G
101933-488EA 600 USD
101933-488 101933-490
Methyl-6-cyano-3-methoxy-2-methylbenzoate
Methyl-6-cyano-3-methoxy-2-methylbenzoate

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
Melting Pt: 73…74 °C
MDL Number: MFCD20921585
CAS Number: 55289-18-4

Order Now

Specification Test Results

Melting point 73-74°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR