Methyl-2-cyano-4-fluoro-5-methoxybenzoate

Supplier: Matrix Scientific

067196-500MG 067196-1G
101933-478EA 803.08 USD
101933-478 101933-480
Methyl-2-cyano-4-fluoro-5-methoxybenzoate
Methyl-2-cyano-4-fluoro-5-methoxybenzoate

Hazard: Irritant

Formula: C₁₀H₈FNO₃
MW: 209.18 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD20482373
CAS Number: 1007455-23-3

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR