Methyl-5-chloro-6-cyanonicotinate

Supplier: Matrix Scientific

067195-500MG 067195-1G
101933-474EA 1923.08 USD
101933-474 101933-476
Methyl-5-chloro-6-cyanonicotinate
Methyl-5-chloro-6-cyanonicotinate

Hazard: Irritant

Formula: C₈H₅ClN₂O₂
Melting Pt: 70…71 °C
MDL Number: MFCD18258701
CAS Number: 398457-04-0

Order Now

Specification Test Results

Melting point 70-71°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR