Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate

Supplier: Matrix Scientific

067194-500MG 067194-1G 067194-5G
101933-468EA 323.08 USD
101933-468 101933-470 101933-472
Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate
Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₇BrN₂O₂
Melting Pt: 70…71 °C
MDL Number: MFCD02179563
CAS Number: 120781-02-4

Order Now

Specification Test Results

Melting point 70-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR