Methyl-5-bromo-2-(methylamino)pyrimidine-4-carboxylate

Supplier: Matrix Scientific

067193-500MG 067193-1G
101933-464EA 600 USD
101933-464 101933-466
Methyl-5-bromo-2-(methylamino)pyrimidine-4-carboxylate
Methyl-5-bromo-2-(methylamino)pyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₇H₈BrN₃O₂
MW: 246.07 g/mol
Melting Pt: 66…68 °C
MDL Number: MFCD20527705
CAS Number: 1009827-04-6

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR