Methyl-2-bromo-5-methoxy-4-methylbenzoate

Supplier: Matrix Scientific

067192-500MG 067192-1G 067192-5G
101933-458EA 212.31 USD
101933-458 101933-460 101933-462
Methyl-2-bromo-5-methoxy-4-methylbenzoate
Methyl-2-bromo-5-methoxy-4-methylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₁BrO₃
MW: 259.11 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD20921581
CAS Number: 87808-28-4

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR