Methyl-4-amino-3-(benzyloxy)benzenecarboxylate

Supplier: Matrix Scientific

067188-500MG 067188-1G
101933-442EA 403.08 USD
101933-442 101933-444
Methyl-4-amino-3-(benzyloxy)benzenecarboxylate
Methyl-4-amino-3-(benzyloxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₅NO₃
MW: 257.29 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD12577089
CAS Number: 475215-88-4

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR