1-Methanesulfonylcyclopropane-1-carboxylic acid

Supplier: Matrix Scientific

067187-500MG 067187-1G
101933-438EA 803.08 USD
101933-438 101933-440
1-Methanesulfonylcyclopropane-1-carboxylic acid
1-Methanesulfonylcyclopropane-1-carboxylic acid

Hazard: Irritant

Formula: C₅H₈O₄S
MW: 164.18 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD16766463
CAS Number: 1249197-58-7

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR