3-Iodo-N-methoxy-N-methyl-5-nitrobenzamide

Supplier: Matrix Scientific

067186-500MG 067186-1G
101933-434EA 803.08 USD
101933-434 101933-436
3-Iodo-N-methoxy-N-methyl-5-nitrobenzamide
3-Iodo-N-methoxy-N-methyl-5-nitrobenzamide

Hazard: Irritant

Formula: C₉H₉IN₂O₄
Melting Pt: 77…78 °C
MDL Number: MFCD20921589
CAS Number: 1211578-13-0

Order Now

Specification Test Results

Melting point 77-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR