3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxamide

Supplier: Matrix Scientific

067181-500MG 067181-1G
101933-418EA 995.38 USD
101933-418 101933-420
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxamide
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxamide

Hazard: Irritant

Formula: C₇H₄F₄N₂O
Melting Pt: 168…169 °C
MDL Number: MFCD16038708
CAS Number: 852062-16-9

Order Now

Specification Test Results

Melting point 168-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR