4-Ethyl-1-(4-nitrophenyl)-1H-1,2,3-triazole

Supplier: Matrix Scientific

067175-500MG 067175-1G
101933-398EA 403.08 USD
101933-398 101933-400
4-Ethyl-1-(4-nitrophenyl)-1H-1,2,3-triazole
4-Ethyl-1-(4-nitrophenyl)-1H-1,2,3-triazole

Hazard: Irritant

Formula: C₁₀H₁₀N₄O₂
Melting Pt: 140…141 °C
MDL Number: MFCD20921608
CAS Number: 1276538-19-2

Order Now

Specification Test Results

Melting point 140-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR