Ethyl-3-methylquinoxaline-2-carboxylate

Supplier: Matrix Scientific

067173-500MG 067173-1G
101933-390EA 600 USD
101933-390 101933-392
Ethyl-3-methylquinoxaline-2-carboxylate
Ethyl-3-methylquinoxaline-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD02182648
CAS Number: 3885-38-9

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR