Ethyl-4,6-dihydroxy-5-iodopyridine-3-carboxylate

Supplier: Matrix Scientific

067170-500MG 067170-1G
101933-378EA 298.46 USD
101933-378 101933-380
Ethyl-4,6-dihydroxy-5-iodopyridine-3-carboxylate
Ethyl-4,6-dihydroxy-5-iodopyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₈INO₄
MW: 309.06 g/mol
Melting Pt: 239…241 °C
MDL Number: MFCD20921606
CAS Number: 1353878-12-2

Order Now

Specification Test Results

Melting point 239-241°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR