Ethyl-4,5-dichloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

Supplier: Matrix Scientific

067169-500MG 067169-1G
101933-374EA 404.62 USD
101933-374 101933-376
Ethyl-4,5-dichloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate
Ethyl-4,5-dichloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

Hazard: Irritant

Formula: C₉H₉Cl₂NO₃
MW: 250.08 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD09031524
CAS Number: 853105-72-3

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR