Ethyl-4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 4-chloro-6-hydroxy-5-nitronicotinate

067168-500MG 067168-1G
101933-370EA 600 USD
101933-370 101933-372
Ethyl-4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate
Ethyl-4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₇ClN₂O₅
MW: 246.603 g/mol
Melting Pt: 176…178 °C
MDL Number: MFCD22989339
CAS Number: 1210835-74-7

Order Now

Specification Test Results

Melting point 176-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR