Ethyl-3-amino-1-methyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Supplier: Matrix Scientific

067165-500MG 067165-1G
101933-358EA 298.46 USD
101933-358 101933-360
Ethyl-3-amino-1-methyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Ethyl-3-amino-1-methyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
MW: 219.25 g/mol
Melting Pt: 128…130 °C
MDL Number: MFCD15529042
CAS Number: 181283-92-1

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR