Ethyl-3-amino-1,6-dimethyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Supplier: Matrix Scientific
067163-500MG
067163-1G
101933-352EA
803.08
USD
101933-352
101933-354
Ethyl-3-amino-1,6-dimethyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Ethyl-3-amino-1,6-dimethyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₄F₃N₃O₂ MW: 301.27 g/mol Melting Pt: 81…83 °C |
MDL Number:
MFCD20921605 CAS Number: 1353877-98-1 |
Specification Test Results
Melting point | 81-83°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...