Ethyl-5-amino-1-(5-chloro-2-pyridinyl)-1H-pyrazole-4-carboxylate

Supplier: Matrix Scientific

067161-500MG 067161-1G
101933-344EA 403.08 USD
101933-344 101933-346
Ethyl-5-amino-1-(5-chloro-2-pyridinyl)-1H-pyrazole-4-carboxylate
Ethyl-5-amino-1-(5-chloro-2-pyridinyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁ClN₄O₂
MW: 266.69 g/mol
Melting Pt: 126…128 °C
MDL Number: MFCD02947352
CAS Number: 104909-58-2

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR