5-(1,3-Dioxan-2-yl)-1H-indazol-3-amine

Supplier: Matrix Scientific

067160-500MG 067160-1G
101933-340EA 403.08 USD
101933-340 101933-342
5-(1,3-Dioxan-2-yl)-1H-indazol-3-amine
5-(1,3-Dioxan-2-yl)-1H-indazol-3-amine

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
Melting Pt: 207…209 °C
MDL Number: MFCD20921613
CAS Number: 218301-24-7

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR