5-(1,3-Dioxan-2-yl)-2-fluorobenzonitrile

Supplier: Matrix Scientific

067159-500MG 067159-1G
101933-336EA 210.77 USD
101933-336 101933-338
5-(1,3-Dioxan-2-yl)-2-fluorobenzonitrile
5-(1,3-Dioxan-2-yl)-2-fluorobenzonitrile

Hazard: Irritant

Formula: C₁₁H₁₀FNO₂
Melting Pt: 45…47 °C
MDL Number: MFCD20921611
CAS Number: 218301-23-6

Order Now

Specification Test Results

Melting point 45-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR