4-{[(2,2-Dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]amino}-5-iodo-2-(trifluoromethyl)benzonitrile

Supplier: Matrix Scientific

067158-500MG 067158-1G
101933-332EA 995.38 USD
101933-332 101933-334
4-{[(2,2-Dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]amino}-5-iodo-2-(trifluoromethyl)benzonitrile
4-{[(2,2-Dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]amino}-5-iodo-2-(trifluoromethyl)benzonitrile

Hazard: Irritant

Formula: C₁₅H₁₀F₃IN₂O₄
MW: 466.16 g/mol
Melting Pt: 225…226 °C (dec.)
MDL Number: MFCD20921595
CAS Number: 1353878-20-2

Order Now

Specification Test Results

Melting point 225-226°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR