4,6-Dichloro-5-(chloromethyl)pyrimidin-2-amine

Supplier: Matrix Scientific

067153-500MG 067153-1G
101933-320EA 515.38 USD
101933-320 101933-322
4,6-Dichloro-5-(chloromethyl)pyrimidin-2-amine
4,6-Dichloro-5-(chloromethyl)pyrimidin-2-amine

Hazard: Irritant

Formula: C₅H₄Cl₃N₃
Melting Pt: 197…201 °C
MDL Number: MFCD19443301
CAS Number: 850554-82-4

Order Now

Specification Test Results

Melting point 197-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR