4-(3,5-Dibromopyridin-2-yl)morpholine

Supplier: Matrix Scientific

067151-500MG 067151-1G 067151-5G
101933-310EA 193.85 USD
101933-310 101933-312 101933-314
4-(3,5-Dibromopyridin-2-yl)morpholine
4-(3,5-Dibromopyridin-2-yl)morpholine

Hazard: Irritant

Formula: C₉H₁₀Br₂N₂O
Melting Pt: 102…104 °C
MDL Number: MFCD20921610
CAS Number: 1259313-58-0

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR