[2-Chloro-4-(1H-1,2,4-triazol-1-yl)phenyl]methanol

Supplier: Matrix Scientific

067149-500MG 067149-1G
101933-302EA 600 USD
101933-302 101933-304
[2-Chloro-4-(1H-1,2,4-triazol-1-yl)phenyl]methanol
[2-Chloro-4-(1H-1,2,4-triazol-1-yl)phenyl]methanol

Hazard: Irritant

Formula: C₉H₈ClN₃O
Melting Pt: 141…144 °C
MDL Number: MFCD20921590
CAS Number: 1353878-01-9

Order Now

Specification Test Results

Melting point 141-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR