4-Chloro-5-nitro-1,3-thiazol-2-ylamine

Supplier: Matrix Scientific

067144-500MG 067144-1G
101933-280EA 803.08 USD
101933-280 101933-282
4-Chloro-5-nitro-1,3-thiazol-2-ylamine
4-Chloro-5-nitro-1,3-thiazol-2-ylamine

Hazard: Irritant

Formula: C₃H₂ClN₃O₂S
Melting Pt: >192 °C (dec.)
MDL Number: MFCD20921569
CAS Number: 127507-89-5

Order Now

Specification Test Results

Melting point >192°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR