4-Chloro-6-methylpyridine-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 4-Chloro-6-methylpicolinic acid

067143-500MG 067143-1G
101933-276EA 538.46 USD
101933-276 101933-278
4-Chloro-6-methylpyridine-2-carboxylic acid
4-Chloro-6-methylpyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 230 °C (dec.)
MDL Number: MFCD07374951
CAS Number: 30235-19-9

Order Now

Specification Test Results

Melting point 230°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR