2-Chloro-5-ethynyl-3-iodo-4-pyridinamine

Supplier: Matrix Scientific

067140-500MG 067140-1G
101933-266EA 803.08 USD
101933-266 101933-268
2-Chloro-5-ethynyl-3-iodo-4-pyridinamine
2-Chloro-5-ethynyl-3-iodo-4-pyridinamine

Hazard: Irritant

Formula: C₇H₄ClIN₂
Melting Pt: 101…102 °C
MDL Number: MFCD20921573
CAS Number: 1353877-92-5

Order Now

Specification Test Results

Melting point 101-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR