5-Bromo-4-methyl-1,3-thiazole-2-carbaldehyde

Supplier: Matrix Scientific

067138-500MG 067138-1G
101933-260EA 1192.31 USD
101933-260 101933-262
5-Bromo-4-methyl-1,3-thiazole-2-carbaldehyde
5-Bromo-4-methyl-1,3-thiazole-2-carbaldehyde

Hazard: Irritant

Formula: C₅H₄BrNOS
Melting Pt: 41…44 °C
MDL Number: MFCD19440535
CAS Number: 95453-56-8

Order Now

Specification Test Results

Melting point 41-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR