2-Bromo-5-methoxy-4-methylbenzoic acid

Supplier: Matrix Scientific

067136-500MG 067136-1G 067136-5G
101933-250EA 146.15 USD
101933-250 101933-252 101933-254
2-Bromo-5-methoxy-4-methylbenzoic acid
2-Bromo-5-methoxy-4-methylbenzoic acid

Hazard: Irritant

Formula: C₉H₉BrO₃
Melting Pt: 189…191 °C
MDL Number: MFCD20921580
CAS Number: 61809-40-3

Order Now

Specification Test Results

Melting point 189-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR