6-Bromo-3-methoxy-2-methylbenzoic acid

Supplier: Matrix Scientific

067135-500MG 067135-1G 067135-5G
101933-244EA 193.85 USD
101933-244 101933-246 101933-248
6-Bromo-3-methoxy-2-methylbenzoic acid
6-Bromo-3-methoxy-2-methylbenzoic acid

Hazard: Irritant

Formula: C₉H₉BrO₃
Melting Pt: 96…98 °C
MDL Number: MFCD16658652
CAS Number: 55289-17-3

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR