5-(Benzyloxy)-6-chloro-4-iodo-2-pyridinecarbaldehyde
Supplier: Matrix Scientific
067130-500MG
067130-1G
101933-224EA
995.38
USD
101933-224
101933-226
5-(Benzyloxy)-6-chloro-4-iodo-2-pyridinecarbaldehyde
5-(Benzyloxy)-6-chloro-4-iodo-2-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₉ClINO₂ Melting Pt: 112…114 °C |
MDL Number:
MFCD20921577 CAS Number: 1353877-99-2 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...