2-(Benzylamino)-5-bromonicotinic acid

Supplier: Matrix Scientific

067128-500MG 067128-1G
101933-216EA 403.08 USD
101933-216 101933-218
2-(Benzylamino)-5-bromonicotinic acid
2-(Benzylamino)-5-bromonicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₁BrN₂O₂
Melting Pt: 231…232 °C
MDL Number: MFCD15503818
CAS Number: 1258846-87-5

Order Now

Specification Test Results

Melting point 231-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR