4-Amino-5-ethynyl-2-(trifluoromethyl)benzenecarbonitrile

Supplier: Matrix Scientific

067124-500MG 067124-1G
101933-206EA 803.08 USD
101933-206 101933-208
4-Amino-5-ethynyl-2-(trifluoromethyl)benzenecarbonitrile
4-Amino-5-ethynyl-2-(trifluoromethyl)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₀H₅F₃N₂
Melting Pt: 122…123 °C
MDL Number: MFCD20921572
CAS Number: 949513-12-6

Order Now

Specification Test Results

Melting point 122-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR