(2-Amino-4,6-dichloropyrimidin-5-yl)methanol

Supplier: Matrix Scientific

067123-500MG 067123-1G
101933-202EA 403.08 USD
101933-202 101933-204
(2-Amino-4,6-dichloropyrimidin-5-yl)methanol
(2-Amino-4,6-dichloropyrimidin-5-yl)methanol

Hazard: Irritant

Formula: C₅H₅Cl₂N₃O
Melting Pt: >200 °C (dec.)
MDL Number: MFCD12756208
CAS Number: 850554-81-3

Order Now

Specification Test Results

Melting point >200°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR