4-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine
067026-500MG
067026-1G
101933-012EA
1192.31
USD
101933-012
101933-014
4-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
4-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
Hazard: Irritant
Formula:
C₇H₅F₃N₄ MW: 202.14 g/mol Melting Pt: 170…172 °C |
MDL Number:
MFCD18261252 CAS Number: 1211579-79-1 |
Specification Test Results
Melting point | 170-172°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...