4-tert-Butylpyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

067025-500MG 067025-1G
101933-008EA 523.08 USD
101933-008 101933-010
4-tert-Butylpyrimidin-2-amine ≥95%
4-tert-Butylpyrimidin-2-amine

Hazard: Irritant

Formula: C₈H₁₃N₃
Melting Pt: 141…144 °C
MDL Number: MFCD01646229
CAS Number: 17321-94-7

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR