1H-Indole-2-carbonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: Indole-2-carbonitrile

067019-500MG 067019-1G
101932-988EA 346.15 USD
101932-988 101932-990
1H-Indole-2-carbonitrile ≥97%
1H-Indole-2-carbonitrile

Hazard: Irritant

Formula: C₉H₆N₂
MW: 142.161 g/mol
Melting Pt: 101 °C
MDL Number: MFCD00187566
CAS Number: 36193-65-4

Order Now

Specification Test Results

Melting point 101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR